Specifications
- Antibody Type:Primary
- Antigen Name:Mucin 6
- Antigen Symbol:MUC6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:200 µl
- Cross Adsorption:No
- Form:Liquid
- Antigen Synonyms:Oligomeric Mucus/Gel-Forming|Mucin 6,Gastric
- Amino Acid Number:2338 - 2439
- UniProtKB:Q6W4X9
- Storage Buffer:0.01 M PBS, pH 7.4, containing 0.05% Proclin-300, 50% glycerol
- Molecular Weight:41 kDa
- Storage Temperature:Store at 2...8 °C for frequent use, −20 °C for 24 months. Avoid repeated freeze-thaw cycles.
- Concentration:0.5 mg/ml (for reference only)
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant MUC6 (Leu2338~Asp2439)
- Purification:Antigen-specific affinity chromatography followed by Protein A affinity chromatography
- Cat. No.:MSPP-PAB489HU1
- Packaging:Vial
Specifications
About this item
Polyclonal Antibody to Mucin 6 (MUC6), derived from recombinant MUC6 (Leu2338~Asp2439), is reactive with Human.
- High specificity, affinity, and purity
- Extensive validation and batch-to-batch consistency
- Diverse range of conjugated and custom options
- Comprehensive validation data and documentation
- All in stock and fast global delivery
- 100% Quality and service satisfaction guarantee
Reid and Harris (1998) used tissue in situ hybridization to study MUC6 gene expression in midtrimester human fetal gut. MUC6 mRNA was detected at low levels in the small intestine between 13 and 23 weeks' gestation. Abundant MUC6 mRNA was detected in gastric neck mucous cells, in contrast with MUC5AC. MUC6 mRNA was present at high levels in pancreatic ductal epithelium throughout the midtrimester., Rousseau et al. (2004) described the domain structure of human MUC6. MUC6 contains an N-terminal signal peptide, followed by several Von Willebrand factor (VWF)-like domains, a large tandem repeat domain, and a C-terminal cystine knot domain.
Type: Primary
Antigen: MUC6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human