Specifications
- Antibody Type:Primary
- Antigen Name:Cat Eye Syndrome Chromosome Region, Candidate 1
- Antigen Symbol:CECR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:200 µl
- Cross Adsorption:No
- Form:Liquid
- Gene ID:51816
- Antigen Synonyms:ADA2|Cat Eye Syndrome Critical Region Protein 1|IDGFL|ADGF
- Amino Acid Number:30 - 511
- UniProtKB:Q9NZK5
- Storage Buffer:PBS, pH 7.4, containing 0.02% NaN₃, 50% glycerol
- Molecular Weight:60 kDa
- Storage Temperature:Store at 2...8 °C for frequent use, −20 °C for 24 months. Avoid repeated freeze-thaw cycles.
- Concentration:1 mg/ml (for reference only)
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant CECR1 (Ile30~Lys511)
- Purification:Antigen-specific affinity chromatography followed by Protein A affinity chromatography
- Cat. No.:MSPP-PAL307HU1
- Packaging:Vial
Specifications
About this item
Polyclonal Antibody to Cat Eye Syndrome Chromosome Region, Candidate 1 (CECR1), derived from recombinant CECR1 (Ile30~Lys511), is reactive with Human/Mouse/Rat.
- High specificity, affinity, and purity
- Extensive validation and batch-to-batch consistency
- Diverse range of conjugated and custom options
- Comprehensive validation data and documentation
- All in stock and fast global delivery
- 100% Quality and service satisfaction guarantee
CECR1 encodes a member of a subfamily of the adenosine deaminase protein family. The encoded protein may act as a growth factor and have adenosine deaminase activity. It may be responsible for some of the phenotypic features associated with cat eye syndrome. Two transcript variants encoding distinct isoforms have been identified for this gene., The deduced protein contains 511 amino acids and shows sequence similarity to insect and mollusk growth factors.Highest expression was in adult heart, lung, lymphoblasts, spleen, and placenta, as well as in fetal lung, liver, and kidney. Expression was weaker in adult pancreas, skeletal muscle, and liver, and faint in adult and fetal brain. In situ hybridization of a 35-day human embryo detected CECR1 mRNA in the outflow tract and atrium of the heart, the VII/VIII cranial nerve ganglion, the notochord, and the placenta.
Type: Primary
Antigen: CECR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat