Specifications
- Antibody Type:Primary
- Antigen Name:A Kinase Anchor Protein 11
- Antigen Symbol:AKAP11
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:200 µl
- Cross Adsorption:No
- Form:Liquid
- Gene ID:11215
- Antigen Synonyms:Protein Kinase A Anchoring Protein 11|A-kinase anchor protein 220 kDa|A Kinase(PRKA)Anchor Protein 11|AKAP220|PRKA11|Protein kinase A-anchoring protein 11
- Amino Acid Number:1650 - 1901
- UniProtKB:Q9UKA4
- Storage Buffer:0.01 M PBS, pH 7.4, containing 0.05% Proclin-300, 50% glycerol
- Molecular Weight:36 kDa
- Storage Temperature:Store at 2...8 °C for frequent use, −20 °C for 24 months. Avoid repeated freeze-thaw cycles.
- Concentration:0.5 mg/ml (for reference only)
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant AKAP11 (Leu1650~Ala1901)
- Purification:Antigen-specific affinity chromatography followed by Protein A affinity chromatography
- Cat. No.:MSPP-PAC197HU1
- Packaging:Vial
Specifications
About this item
Polyclonal Antibody to A Kinase Anchor Protein 11 (AKAP11), derived from recombinant AKAP11 (Leu1650~Ala1901), is reactive with Human.
- High specificity, affinity, and purity
- Extensive validation and batch-to-batch consistency
- Diverse range of conjugated and custom options
- Comprehensive validation data and documentation
- All in stock and fast global delivery
- 100% Quality and service satisfaction guarantee
A-kinase anchor protein 11 is expressed at high levels throughout spermatogenesis and in mature sperm. It binds the RI and RII subunits of PKA in testis. It may serve a function in cell cycle control of both somatic cells and germ cells in addition to its putative role in spermatogenesis and sperm function. The AKAP11 protein has 1129 amino acids and contains a PKA-binding region and a peroxisome targeting motif. Immunohistochemical analysis of rat testis cells showed that AKAP220 and a proportion of RII PKA are colocalized in microbodies that appear to be a subset of peroxisomes. By RT-PCR analysis, Ishikawa et al. (1998) detected expression of human AKAP11 in heart, brain, lung, liver, kidney, testis, and ovary, with weaker expression in skeletal muscle, pancreas, and spleen.
Type: Primary
Antigen: AKAP11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human