Anti-PKM2 Rabbit Polyclonal Antibody
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Polyclonal Antibody to Pyruvate kinase isozymes M2 (PKM2), derived from recombinant PKM2 (Lys141~Ala248), is reactive with Human.
- High specificity, affinity, and purity
- Extensive validation and batch-to-batch consistency
- Diverse range of conjugated and custom options
- Comprehensive validation data and documentation
- All in stock and fast global delivery
- 100% Quality and service satisfaction guarantee
Pyruvate Kinase M2 (M2-PK) is strongly overexpressed in a metabolic state specific for a variety of tumor cells. Recent reports indicate that Tumor M2-PK is a marker for renal cell carcinoma and. lung cancer. It has also been investigated in. pancreatic cancer and gastric cancer with promising results., M2-PK is encoded by the gene PKM2 which does also encode for the pyruvate kinase isoenzyme type M1 (PKM1 or M1-PK). The M-gene consists of 12 exons and 11 introns. PKM1 and PKM2 are different splicing products of the M-gene (exon 9 for PKM1 and exon 10 for PKM2) and solely differ in 23 amino acids within a 56 amino acid stretch (aa 378-434) at their carboxy terminus.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.
Type: Primary
Antigen: PKM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Documents
Cloud-Clone Quality Guarantee
Additional Product Details
Specifications
- Size:200 µl
- Packaging:Vial
- Clonality:Polyclonal
- Molecular Weight:13 kDa
- Form:Liquid
- ImmunoFluorescence:Yes
- UniProtKB:P14618
- Antibody Type:Primary
- Gene ID:5315
- Immunogen:Recombinant PKM2 (Lys141~Ala248)
- Amino Acid Number:141 - 248
- Storage Temperature:Store at 2...8 °C for frequent use, −20 °C for 24 months. Avoid repeated freeze-thaw cycles.
- Cross Adsorption:No
- Purification:Antigen-specific affinity chromatography followed by Protein A affinity chromatography
- Storage Buffer:0.01 M PBS, pH 7.4, containing 0.05% Proclin-300, 50% glycerol
- Isotype:IgG
- Host:Rabbit
- Concentration:0.5 mg/ml (for reference only)
- Cat. No.:MSPP-PAA588HU1
- Reactivity:Human
- Western Blot:Yes
- Shipping Temperature:Shipped on blue ice at 4 °C
- Antigen Symbol:PKM2
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen Synonyms:PKM1|M2PK|THBP1|TCB|CTHBP|PKM|Pyruvate Kinase, Muscle|Thyroid hormone-binding protein 1|PK3|M2-PK|OIP3|Cytosolic thyroid hormone-binding protein|Pyruvate kinase muscle isozyme
- Antigen Name:Pyruvate kinase isozymes M2
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



