- Antibody Type:Primary
- Antigen Name:Myostatin
- Antigen Symbol:MSTN
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Bovine,Rat,Mouse
- Western Blot:Yes
- Size:200 µl
- Cross Adsorption:No
- Form:Liquid
- Gene ID:281187
- Antigen Synonyms:GDF8|Growth Differentiation Factor 8
- Amino Acid Number:267 - 375
- UniProtKB:O18836
- Storage Buffer:PBS, pH 7.4, containing 0.02% NaN₃, 50% glycerol
- Molecular Weight:16 kDa
- Storage Temperature:Store at 2...8 °C for frequent use, −20 °C for 24 months. Avoid repeated freeze-thaw cycles.
- Concentration:0.5 mg/ml (for reference only)
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant MSTN (Asp267~Ser375)
- Purification:Antigen-specific affinity chromatography followed by Protein A affinity chromatography
- Cat. No.:MSPP-PAB653BO1
- Packaging:Vial
Polyclonal Antibody to Myostatin (MSTN), derived from recombinant MSTN (Asp267~Ser375), is reactive with Bovine/Human/Mouse/Rat.
- High specificity, affinity, and purity
- Extensive validation and batch-to-batch consistency
- Diverse range of conjugated and custom options
- Comprehensive validation data and documentation
- All in stock and fast global delivery
- 100% Quality and service satisfaction guarantee
Myostatin is a growth factor that limits muscle tissue growth, i.e. higher concentrations of myostatin in the body may cause the individual to have less developed muscles. The myostatin protein is produced primarily in skeletal muscle cells, circulates in the blood and acts on muscle tissue, apparently by slowing down the development of muscle stem cells. The precise mechanism remains unknown. Its functions in non-mammalian vertebrates appear to be somewhat conserved as muscle-specific actions have been demonstrated in birds. However, it is produced in many different fish tissues, suggesting that it may regulate more than just muscle mass in these vertebrates. Human Myostatin consists of two identical subunits, each consisting of 110 amino acid residues. Its total molecular weight is 25.0 kDa.
Type: Primary
Antigen: MSTN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine;Human;Mouse;Rat