- Antibody Type:Primary
- Antigen Name:Gastrin
- Antigen Symbol:GT
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Pig,Human,Guinea pig,Rat,Mouse
- Western Blot:Yes
- Size:200 µl
- Cross Adsorption:No
- Form:Liquid
- Gene ID:2520
- Antigen Synonyms:Gastrin component III|Big gastrin|GAS|GAST|Gastrin component I|Gastrin component II
- Amino Acid Number:1 - 101
- UniProtKB:P01350
- Storage Buffer:PBS, pH 7.4, containing 0.02% NaN₃, 50% glycerol
- Molecular Weight:42 kDa
- Storage Temperature:Store at 2...8 °C for frequent use, −20 °C for 24 months. Avoid repeated freeze-thaw cycles.
- Concentration:0.5 mg/ml (for reference only)
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant GT (Met1~Asn101)
- Purification:Antigen-specific affinity chromatography followed by Protein A affinity chromatography
- Cat. No.:MSPP-PAB224HU1
- Packaging:Vial
Polyclonal Antibody to Gastrin (GT), derived from recombinant GT (Met1~Asn101), is reactive with Human/Mouse/Rat/Guinea pig/Pig.
- High specificity, affinity, and purity
- Extensive validation and batch-to-batch consistency
- Diverse range of conjugated and custom options
- Comprehensive validation data and documentation
- All in stock and fast global delivery
- 100% Quality and service satisfaction guarantee
Gastrin is a major physiological regulator of gastric acid secretion. It also has an important trophic or growth-promoting influence on the gastric mucosa. Gastrin is synthesized in G cells, which are located in gastric pits, primarily in the antrum region of the stomach and binds receptors found predominantly on parietal and enterochromaffin-like cells.In humans, gastrin is a hormone that stimulates secretion of gastric acid by the parietal cells of the stomach., It is released by G cells in the stomach and duodenum.Gastrin is a linear peptide hormone produced by G cells of the duodenum and in the pyloric antrum of the stomach. In the Zollinger-Ellison syndrome, gastrin is produced at excessive levels, often by a gastrinoma of the antrum or the pancreas.
Type: Primary
Antigen: GT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat;Guinea pig;Pig