- Antibody Type:Primary
- Antigen Name:Sialic Acid Binding Ig Like Lectin 10
- Antigen Symbol:SIGLEC10
- Clonality:Monoclonal
- Clone:C5
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:200 µl
- Cross Adsorption:No
- Form:Liquid
- Gene ID:89790
- Antigen Synonyms:Siglec-Like Gene 2|PRO940|SLG2|Sialic Acid Binding Ig-Like Lectin 10 Ig-Like Lectin 7
- Amino Acid Number:571 - 697
- UniProtKB:Q96LC7
- Storage Buffer:0.01 M PBS, pH 7.4, containing 0.05% Proclin-300, 50% glycerol
- Molecular Weight:17 kDa
- Storage Temperature:Store at 2...8 °C for frequent use, −20 °C for 24 months. Avoid repeated freeze-thaw cycles.
- Concentration:1 mg/ml (for reference only)
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant SIGLEC10 (Met571~Gln697)
- Purification:Protein A + Protein G affinity chromatography
- Cat. No.:MSPP-MD921HU21
- Packaging:Vial
Monoclonal Antibody to Sialic Acid Binding Ig Like Lectin 10 (SIGLEC10), derived from recombinant SIGLEC10 (Met571~Gln697), is reactive with Human.
- High specificity, affinity, and purity
- Extensive validation and batch-to-batch consistency
- Diverse range of conjugated and custom options
- Comprehensive validation data and documentation
- All in stock and fast global delivery
- 100% Quality and service satisfaction guarantee
By searching an EST database for sequences resembling CD33 (SIGLEC3) and screening a spleen cDNA library, Munday et al. (2001) isolated a cDNA encoding SIGLEC10. Sequence analysis predicted that the 697-amino acid SIGLEC10 protein contains a signal peptide, an N-terminal V-set Ig-like domain and 4 C2-set Ig-like domains, 5 potential N-linked glycosylation sites, a transmembrane region, and a 126-residue cytoplasmic tail with 3 putative ITIMs. Northern blot analysis detected a major 3.0-kb SIGLEC10 transcript, with highest levels in spleen, lymph node, blood leukocytes, and appendix. Little or no expression was observed in pancreas, thyroid, and testis. Flow cytometric analysis demonstrated eosinophil-specific expression of SIGLEC10, but at a lower level than that of SIGLEC8.
Type: Primary
Antigen: SIGLEC10
Clonality: Monoclonal
Clone: C5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human