Specifications
- Antibody Type:Primary
- Antigen Name:Cyclic Adenosine Monophosphate
- Antigen Symbol:cAMP
- Clonality:Monoclonal
- Clone:1-C7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Reactivity:Multiple
- Size:200 µl
- Cross Adsorption:No
- Form:Liquid
- Antigen Synonyms:Adenosine Cyclophosphate|c-AMP|3'-5'-Cyclic Adenosine Monophosphate
- Storage Buffer:PBS, pH 7.4, containing 0.02% NaN₃, 50% glycerol
- Molecular Weight:329.2 Da
- Storage Temperature:Store at 2...8 °C for frequent use, −20 °C for 24 months. Avoid repeated freeze-thaw cycles.
- Concentration:1 mg/ml (for reference only)
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:BSA conjugated cAMP
- Purification:Protein A + Protein G affinity chromatography
- Cat. No.:MSPP-MAA03GE01
- Packaging:Vial
Specifications
About this item
Monoclonal Antibody to Cyclic Adenosine Monophosphate (cAMP), derived from BSA conjugated cAMP, is reactive with General species.
- High specificity, affinity, and purity
- Extensive validation and batch-to-batch consistency
- Diverse range of conjugated and custom options
- Comprehensive validation data and documentation
- All in stock and fast global delivery
- 100% Quality and service satisfaction guarantee
Cyclic adenosine monophosphate is a second messenger important in many biological processes. cAMP is derived from adenosine triphosphate (ATP) and used for intracellular signal transduction in many different organisms, conveying the cAMP-dependent pathway. cAMP is synthesised from ATP by adenylyl cyclase located at the cell membranes., Adenylyl cyclase is activated by a range of signaling molecules through the activation of adenylyl cyclase stimulatory G (Gs)-coupled receptors and inhibited by agonists of adenylyl cyclase inhibitory G (Gi)-protein-coupled receptors. Liver adenylyl cyclase responds more strongly to glucagon, and muscle adenylyl cyclase responds more strongly to adrenaline. cAMP is a second messenger, used for intracellular signal transduction, such as transferring the effects of hormones like glucagon and adrenaline, which cannot pass through the cell membrane.
Type: Primary
Antigen: cAMP
Clonality: Monoclonal
Clone: 1-C7
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1 kappa
Reactivity: Multiple