- Antibody Type:Primary
- Antigen Name:Glyceraldehyde-3-Phosphate Dehydrogenase
- Antigen Symbol:GAPDH
- Clonality:Monoclonal
- Clone:E4
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Reactivity:Pig,Rabbit,Sheep,Goat,Chicken,Human,Guinea pig,Rat,Mouse
- Western Blot:Yes
- Size:200 µl
- Cross Adsorption:No
- Form:Liquid
- Gene ID:2597
- Antigen Synonyms:G3PD|GAPD|Peptidyl-cysteine S-nitrosylase GAPDH
- Amino Acid Number:2 - 148
- UniProtKB:P04406
- Storage Buffer:0.01 M PBS, pH 7.4, containing 0.05% Proclin-300, 50% glycerol
- Molecular Weight:18 kDa
- Storage Temperature:Store at 2...8 °C for frequent use, −20 °C for 24 months. Avoid repeated freeze-thaw cycles.
- Concentration:1 mg/ml (for reference only)
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant GAPDH (Gly2~Ser148)
- Purification:Affinity chromatography
- Cat. No.:MSPP-MB932HU21
- Packaging:Vial
Monoclonal Antibody to Glyceraldehyde-3-Phosphate Dehydrogenase (GAPDH), derived from recombinant GAPDH (Gly2~Ser148), is reactive with Human/Mouse/Rat/Guinea pig/Rabbit/Pig/Goat/Sheep/Chicken.
- High specificity, affinity, and purity
- Extensive validation and batch-to-batch consistency
- Diverse range of conjugated and custom options
- Comprehensive validation data and documentation
- All in stock and fast global delivery
- 100% Quality and service satisfaction guarantee
Glyceraldehyde 3-phosphate dehydrogenase is an enzyme that catalyzes the sixth step of glycolysis and thus serves to break down glucose for energy and carbon molecules. In addition to this long established metabolic function, GAPDH has recently been implicated in several non-metabolic processes, including transcription activation, initiation of apoptosis, and ER to Golgi vesicle shuttling. Glyceraldehyde 3-phosphate dehydrogenase (GAPDH) catalyses the conversion of glyceraldehyde 3-phosphate as the name indicates. This is the 6th step of the breakdown of glucose (glycolysis), an important pathway of energy and carbon molecule supply located in the cytosol of eukaryotic cells. Glyceraldehyde 3-phosphate is converted to D-glycerate 1,3-bisphosphate in two coupled steps.
Type: Primary
Antigen: GAPDH
Clonality: Monoclonal
Clone: E4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human;Mouse;Rat;Guinea pig;Rabbit;Pig;Goat;Sheep;Chicken