- Antibody Type:Primary
- Antigen Name:Alanine Glyoxylate Aminotransferase 2
- Antigen Symbol:AGXT2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:200 µl
- Cross Adsorption:No
- Form:Liquid
- Gene ID:268782
- Antigen Synonyms:(R)-3-amino-2-methylpropionate--pyruvate transaminase|Beta-Alanine-Pyruvate Aminotransferase|AGT2
- Amino Acid Number:226 - 513
- UniProtKB:Q3UEG6
- Storage Buffer:PBS, pH 7.4, containing 0.02% NaN₃, 50% glycerol
- Molecular Weight:35 kDa
- Storage Temperature:Store at 2...8 °C for frequent use, −20 °C for 24 months. Avoid repeated freeze-thaw cycles.
- Concentration:0.5 mg/ml (for reference only)
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant AGXT2 (Met226~Lys513)
- Purification:Antigen-specific affinity chromatography followed by Protein A affinity chromatography
- Cat. No.:MSPP-PAD855MU1
- Packaging:Vial
Polyclonal Antibody to Alanine Glyoxylate Aminotransferase 2 (AGXT2), derived from recombinant AGXT2 (Met226~Lys513), is reactive with Mouse/Human/Rat.
- High specificity, affinity, and purity
- Extensive validation and batch-to-batch consistency
- Diverse range of conjugated and custom options
- Comprehensive validation data and documentation
- All in stock and fast global delivery
- 100% Quality and service satisfaction guarantee
AGXT2 encodes the mitochondrial alanine-glyoxylate aminotransferase, which catalyzes the conversion of glyoxylate to glycine using L-alanine as the amino donor.The deduced 512-amino acid rat Agxt2 enzyme has a predicted 39-amino acid precursor leader sequence and has a calculated molecular mass of 57.15 kDa. Rat Agxt2 shares 29.2% identity with rat ornithine aminotransferase (OAT1) but has little similarity to rat mitochondrial Agt1 or to human peroxisomal Agt1 (AGXT).Enzyme activity assay detected alanine-glyoxylate aminotransferase and glycolate reductase activity in purified mitochondria of liver carcinoma cells, and beta-ALAAT II activity was enriched in mitochondria, suggesting that human hepatocyte mitochondria possess AGXT2 activity.
Type: Primary
Antigen: AGXT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Human;Rat