- Antibody Type:Primary
- Antigen Name:Cathelicidin Antimicrobial Peptide
- Antigen Symbol:CAMP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Mouse
- Western Blot:Yes
- Size:200 µl
- Cross Adsorption:No
- Form:Liquid
- Gene ID:12796
- Antigen Synonyms:CAP18|LL37|FALL-39 peptide antibiotic|18 kDa Cationic Antimicrobial Protein|HSD26|Antibacterial protein FALL-39|FALL39
- Amino Acid Number:31 - 172
- UniProtKB:P51437
- Storage Buffer:0.01 M PBS, pH 7.4, containing 0.05% Proclin-300, 50% glycerol
- Molecular Weight:48 kDa
- Storage Temperature:Store at 2...8 °C for frequent use, −20 °C for 24 months. Avoid repeated freeze-thaw cycles.
- Concentration:0.5 mg/ml (for reference only)
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant CAMP (Ser31~Glu172)
- Purification:Antigen-specific affinity chromatography followed by Protein A affinity chromatography
- Cat. No.:MSPP-PAC419MU1
- Packaging:Vial
Polyclonal Antibody to Cathelicidin Antimicrobial Peptide (CAMP), derived from recombinant CAMP (Ser31~Glu172), is reactive with Mouse.
- High specificity, affinity, and purity
- Extensive validation and batch-to-batch consistency
- Diverse range of conjugated and custom options
- Comprehensive validation data and documentation
- All in stock and fast global delivery
- 100% Quality and service satisfaction guarantee
Cathelicidin antimicrobial peptide is a family of polypeptides found in lysosomes in polymorphonuclear leukocytes (PMNs).Members of the cathelicidin family of antimicrobial polypeptides are characterized by a highly conserved region (cathelin domain) and a highly variable cathelicidin peptide domain. Cathelicidin peptides have been isolated from many different species of mammals. Cathelicidins were originally found in neutrophils but have since been found in many other cells including epithelial cells and macrophages activated by bacteria, viruses, fungi, or the hormone 1,25-D. The cathelicidin family shares primary sequence homology with the cathepsin family of cysteine proteinase inhibitors, although amino acid residues thought to be important in such protease inhibition are usually lacking.
Type: Primary
Antigen: CAMP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse