Specifications
- Antibody Type:Primary
- Antigen Name:Matrix Remodelling Associated Protein 5
- Antigen Symbol:MXRA5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:200 µl
- Cross Adsorption:No
- Form:Liquid
- Gene ID:25878
- Antigen Synonyms:Adlican|Adhesion protein with leucine-rich repeats and immunoglobulin domains related to perlecan
- Amino Acid Number:2542 - 2828
- UniProtKB:Q9NR99
- Storage Buffer:0.01 M PBS, pH 7.4, containing 0.05% Proclin-300, 50% glycerol
- Molecular Weight:35 kDa
- Storage Temperature:Store at 2...8 °C for frequent use, −20 °C for 24 months. Avoid repeated freeze-thaw cycles.
- Concentration:0.5 mg/ml (for reference only)
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant MXRA5 (Pro2542~Phe2828)
- Purification:Antigen-specific affinity chromatography followed by Protein A affinity chromatography
- Cat. No.:MSPP-PAP859HU1
- Packaging:Vial
Specifications
About this item
Polyclonal Antibody to Matrix Remodelling Associated Protein 5 (MXRA5), derived from recombinant MXRA5 (Pro2542~Phe2828), is reactive with Human.
- High specificity, affinity, and purity
- Extensive validation and batch-to-batch consistency
- Diverse range of conjugated and custom options
- Comprehensive validation data and documentation
- All in stock and fast global delivery
- 100% Quality and service satisfaction guarantee
Adlican is one of the matrix-remodelling associated proteins. This protein contains 7 leucine-rich repeats and 12 immunoglobulin-like C2-type domains related to perlecan. This gene has a pseudogene on chromosome Y., In order to discover global gene expression patterns characterizing subgroups of colon cancer, microarrays were hybridized to labeled RNAs obtained from seventeen colonic specimens. Using a hierarchical agglomerative method, the samples grouped naturally into two major clusters, in perfect concordance with pathological reports. Using a variant of the unpaired t-test, selected genes were ordered according to an index of importance. RT-PCR was performed on the gro1, B-factor, adlican, and endothelin converting enzyme-1 genes and confirmed microarray findings.
Type: Primary
Antigen: MXRA5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human