- Antibody Type:Primary
- Antigen Name:Complexin 2
- Antigen Symbol:CPLX2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:200 µl
- Cross Adsorption:No
- Form:Liquid
- Gene ID:10814
- Antigen Synonyms:CPX2|921-L|CPX-2|Synaphin-1
- Amino Acid Number:2 - 134
- UniProtKB:Q6PUV4
- Storage Buffer:0.01 M PBS, pH 7.4, containing 0.05% Proclin-300, 50% glycerol
- Molecular Weight:19 kDa
- Storage Temperature:Store at 2...8 °C for frequent use, –20 ℃ for 24 months. Avoid repeated freeze-thaw cycles.
- Concentration:0.5 mg/ml (for reference only)
- Shipping Temperature:Shipped on blue ice at 4 °C.
- Immunogen:Recombinant CPLX2 (Asp2~Lys134)
- Purification:Antigen-specific affinity chromatography followed by Protein A affinity chromatography
- Cat. No.:MSPP-PAF332HU1
- Packaging:Vial
Polyclonal Antibody to Complexin 2 (CPLX2), derived from recombinant CPLX2 (Asp2~Lys134), is reactive with Human/Mouse/Rat.
- High specificity, affinity, and purity
- Extensive validation and batch-to-batch consistency
- Diverse range of conjugated and custom options
- Comprehensive validation data and documentation
- All in stock and fast global delivery
- 100% Quality and service satisfaction guarantee
Proteins encoded by the complexin/synaphin gene family are cytosolic proteins that function in synaptic vesicle exocytosis. These proteins bind syntaxin, part of the SNAP receptor. The protein product of this gene binds to the SNAP receptor complex and disrupts it, allowing transmitter release. Two transcript variants encoding the same protein have been found for this gene. Sequence analysis determined that the rat Cplx1 and Cplx2 genes encode 134-amino acid, highly charged proteins that share 84% amino acid identity. Asp, glu, lys, and arg account for 44 to 47% of the complexin protein residues. The amino acid sequences, but not the nucleotide sequences, of mouse, rat, and human CPLX2 are 100% identical.
Type: Primary
Antigen: CPLX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat