- Antibody Type:Primary
- Antigen Name:Trypsin
- Antigen Symbol:TRY
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Pig,Human,Rat
- Western Blot:Yes
- Size:200 µl
- Cross Adsorption:No
- Form:Liquid
- Amino Acid Number:9 - 231
- UniProtKB:P00761
- Storage Buffer:0.01 M PBS, pH 7.4, containing 0.05% Proclin-300, 50% glycerol
- Molecular Weight:26 kDa
- Storage Temperature:Store at 2...8 °C for frequent use, –20 ℃ for 24 months. Avoid repeated freeze-thaw cycles.
- Concentration:0.5 mg/ml (for reference only)
- Shipping Temperature:Shipped on blue ice at 4 °C.
- Immunogen:Recombinant TRY (Ile9~Asn231)
- Purification:Antigen-specific affinity chromatography followed by Protein A affinity chromatography
- Cat. No.:MSPP-PAA250PO1
- Packaging:Vial
Polyclonal Antibody to Trypsin (TRY), derived from recombinant TRY (Ile9~Asn231), is reactive with Pig/Human/Rat.
- High specificity, affinity, and purity
- Extensive validation and batch-to-batch consistency
- Diverse range of conjugated and custom options
- Comprehensive validation data and documentation
- All in stock and fast global delivery
- 100% Quality and service satisfaction guarantee
Trypsin is produced in the pancreas as the inactive proenzyme trypsinogen. Trypsin predominantly cleaves peptide chains at the carboxyl side of the amino acids lysine or arginine, except when either is followed by proline. It is used for numerous biotechnological processes. The process is commonly referred to as trypsin proteolysis or trypsinisation and proteins that have been digested/treated with trypsin are said to have been trypsinized. Trypsin is secreted into the duodenum, where it acts to hydrolyse peptides into their smaller building blocks, namely amino acids (these peptides are the result of the enzyme pepsin breaking down the proteins in the stomach). This is necessary for the uptake of protein in the food as though peptides are smaller than proteins, they are still too big to be absorbed through the lining of the ileum. Trypsin catalyses the hydrolysis of peptide bonds.
Type: Primary
Antigen: TRY
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Pig, Human, Rat