Specifications
- Antibody Type:Primary
- Antigen Name:Interferon Alpha/Beta Receptor 2
- Antigen Symbol:IFNa/bR2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Pig,Human,Rat,Mouse
- Western Blot:Yes
- Size:200 µl
- Cross Adsorption:No
- Form:Liquid
- Gene ID:3455
- Antigen Synonyms:IFNARB|Type I interferon receptor 2|IFN-R|IFN-alpha-REC|IFNABR|IFNAR2|Interferon(Alpha,Beta And Omega)Receptor Beta Chain|Interferon alpha binding protein
- Amino Acid Number:27 - 243
- UniProtKB:P48551
- Storage Buffer:0.01 M PBS, pH 7.4, containing 0.05% Proclin-300, 50% glycerol
- Molecular Weight:29 kDa
- Storage Temperature:Store at 2...8 °C for frequent use, –20 ℃ for 24 months. Avoid repeated freeze-thaw cycles.
- Concentration:0.5 mg/ml (for reference only)
- Shipping Temperature:Shipped on blue ice at 4 °C.
- Immunogen:Recombinant IFNa/bR2 (Ile27~Lys243)
- Purification:Antigen-specific affinity chromatography followed by Protein A affinity chromatography
- Cat. No.:MSPP-PAE171HU1
- Packaging:Vial
Specifications
About this item
Polyclonal Antibody to Interferon Alpha/Beta Receptor 2 (IFNa/bR2), derived from recombinant IFNa/bR2 (Ile27~Lys243), is reactive with Human/Mouse/Rat/Pig.
- High specificity, affinity, and purity
- Extensive validation and batch-to-batch consistency
- Diverse range of conjugated and custom options
- Comprehensive validation data and documentation
- All in stock and fast global delivery
- 100% Quality and service satisfaction guarantee
IFNAR2 was identified by several groups, including Novick et al. (1994), Lutfalla et al. (1995), and Domanski et al. (1995). Lutfalla et al. (1995) reported the structure of the IFNAR2 gene and showed that it is located at chromosome 21q22.1 in a cluster with the class II cytokine receptor genes CRFB4 (IL10RB), IFNAR1, and AF1. They found evidence for alternative splicing of the IFNAR2 gene and showed that one of the transcripts, ifnar2-2, could functionally complement the IFN-alpha/beta receptor-defective mutant U5A cell line. Raz et al. (1995) independently reported using human-hamster somatic cell hybrids to map IFNAR2 to 21q22.1. There was no significant difference in RNA expression of the F8 and S8 IFNAR2 variants, but FACS analysis demonstrated higher cell surface expression of the F8 variant.
Type: Primary
Antigen: IFNa/bR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Pig