- Antibody Type:Primary
- Antigen Name:Follistatin Like Protein 3
- Antigen Symbol:FSTL3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Mouse
- Western Blot:Yes
- Size:200 µl
- Cross Adsorption:No
- Form:Liquid
- Gene ID:83554
- Antigen Synonyms:FSRP|FLRG|Follistatin-related gene protein|Follistatin-Related Protein,Secreted Glycoprotein
- Amino Acid Number:24 - 256
- UniProtKB:Q9EQC7
- Storage Buffer:0.01 M PBS, pH 7.4, containing 0.05% Proclin-300, 50% glycerol
- Molecular Weight:29 kDa
- Storage Temperature:Store at 2...8 °C for frequent use, –20 ℃ for 24 months. Avoid repeated freeze-thaw cycles.
- Concentration:0.5 mg/ml (for reference only)
- Shipping Temperature:Shipped on blue ice at 4 °C.
- Immunogen:Recombinant FSTL3 (Val24~Val256)
- Purification:Antigen-specific affinity chromatography followed by Protein A affinity chromatography
- Cat. No.:MSPP-PAJ084MU1
- Packaging:Vial
Polyclonal Antibody to Follistatin Like Protein 3 (FSTL3), derived from recombinant FSTL3 (Val24~Val256), is reactive with Mouse.
- High specificity, affinity, and purity
- Extensive validation and batch-to-batch consistency
- Diverse range of conjugated and custom options
- Comprehensive validation data and documentation
- All in stock and fast global delivery
- 100% Quality and service satisfaction guarantee
Follistatin-like 3 is a secreted glycoprotein of the follistatin-module-protein family. It may have a role in leukemogenesis. The FSTL3 cDNA, which they called FLRG (follistatin-related gene), encodes a deduced 263-amino acid protein that shares 85% amino acid identity with mouse Fstl3. It contains 2 cysteine-rich repeats that are closely related to the class 1-1 follistatin modules present in other follistatin family members. Northern blot analysis demonstrated expression of a 2.5-Kb transcript in placenta, amniocytes, chorionic villosities, and bone marrow stromal cells, with weaker expression in fibroblasts, keratinocytes, and endothelial cells. A 1.3-Kb transcript was also detected in some human tissues and was thought to result from use of an alternate polyadenylation site.
Type: Primary
Antigen: FSTL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse