Specifications
- Antibody Type:Primary
- Antigen Name:Integrin Beta 1
- Antigen Symbol:ITGb1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:200 µl
- Cross Adsorption:No
- Form:Liquid
- Gene ID:3688
- Antigen Synonyms:FNRB|Glycoprotein IIa|VLAB|VLA-4 subunit beta|CD29|MDF2|MSK12|GPIIA|ITG-B1|Antigen CD29 Includes MDF2|Fibronectin Receptor Beta Polypeptide
- Amino Acid Number:140 - 383
- UniProtKB:P05556
- Storage Buffer:0.01 M PBS, pH 7.4, containing 0.05% Proclin-300, 50% glycerol
- Molecular Weight:57 kDa
- Storage Temperature:Store at 2...8 °C for frequent use, –20 ℃ for 24 months. Avoid repeated freeze-thaw cycles.
- Concentration:0.5 mg/ml (for reference only)
- Shipping Temperature:Shipped on blue ice at 4 °C.
- Immunogen:Recombinant ITGb1 (Asp140~Ile383)
- Purification:Antigen-specific affinity chromatography followed by Protein A affinity chromatography
- Cat. No.:MSPP-PAB042HU1
- Packaging:Vial
Specifications
About this item
Polyclonal Antibody to Integrin Beta 1 (ITGb1), derived from recombinant ITGb1 (Asp140~Ile383), is reactive with Human/Mouse/Rat.
- High specificity, affinity, and purity
- Extensive validation and batch-to-batch consistency
- Diverse range of conjugated and custom options
- Comprehensive validation data and documentation
- All in stock and fast global delivery
- 100% Quality and service satisfaction guarantee
Integrin, beta 1 (fibronectin receptor, beta polypeptide, antigen CD29 includes MDF2, MSK12), also known as ITGB1 or CD29, is a human gene. CD29 is an integrin unit associated with very late antigen receptors. Integrins are heterodimeric proteins made up of alpha and beta subunits. At least 18 alpha and 8 beta subunits have been described in mammals. Integrin family members are membrane receptors involved in cell adhesion and recognition in a variety of processes including embryogenesis, hemostasis, tissue repair, immune response and metatastatic diffusion of tumor cells. The protein encoded by this gene is a beta subunit. Six alternatively spliced variants have been found for this gene which encode five proteins with alternate carboxy termini.
Type: Primary
Antigen: ITGb1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat