Specifications
- Antibody Type:Primary
- Antigen Name:Triiodothyronine
- Antigen Symbol:T3
- Clonality:Monoclonal
- Clone:E5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2b kappa
- Reactivity:Multiple
- Size:200 µl
- Cross Adsorption:No
- Form:Liquid
- Antigen Synonyms:3,3',5-Triiodo-L-Thyronine|Cytomel|Liothyronine|Tertroxin
- Storage Buffer:0.01 M PBS, pH 7.4, containing 0.05% Proclin-300, 50% glycerol
- Molecular Weight:651.0 Da
- Storage Temperature:Store at 2...8 °C for frequent use, –20 ℃ for 24 months. Avoid repeated freeze-thaw cycles.
- Concentration:1 mg/ml (for reference only)
- Shipping Temperature:Shipped on blue ice at 4 °C.
- Immunogen:BSA conjugated T3
- Purification:Protein A + Protein G affinity chromatography
- Cat. No.:MSPP-MAA453GE2
- Packaging:Vial
Specifications
About this item
Monoclonal Antibody to Triiodothyronine (T3), derived from BSA conjugated T3, is reactive with General species.
- High specificity, affinity, and purity
- Extensive validation and batch-to-batch consistency
- Diverse range of conjugated and custom options
- Comprehensive validation data and documentation
- All in stock and fast global delivery
- 100% Quality and service satisfaction guarantee
Under normal physiological conditions, T3 represents approximately 5 percent of the thyroid hormones in serum. Although present in lower concentration, T3 has a greater intrinsic metabolic activity, faster turnover and larger volume of distribution than circulating T4. Reports that thyrotoxicosis may be caused by abnormally high concentrations of T3 rather than T4 have reinforced the importance of T3 measurements. In addition, T3 determination is an important tool for monitoring hypothyroid patients receiving sodium liothyronine therapy. Unlike 'T3 Uptake' tests, which estimate the saturation of thyroid hormone binding proteins, total T3 analysis measures circulating levels of triiodothyronine. Most reports indicate that T3 levels distinguish clearly between euthyroid and hyperthyroid subjects, but provide a less clear-cut separation between hypothyroid and euthyroid subjects. Numerous conditions unrelated to thyroid disease may cause abnormal T3 values.
Type: Primary
Antigen: T3
Clonality: Monoclonal
Clone: E5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Multiple