Anti-Cor Mouse Monoclonal Antibody [clone: C2]
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Monoclonal Antibody to Cortisol (Cor), derived from OVA conjugated Cor, is reactive with General species.
- High specificity, affinity, and purity
- Extensive validation and batch-to-batch consistency
- Diverse range of conjugated and custom options
- Comprehensive validation data and documentation
- All in stock and fast global delivery
- 100% Quality and service satisfaction guarantee
Cortisol is the major glucocorticoid produced and secreted by the adrenal cortex. It affects the metabolism of protein, fat and carbohydrates; the maintenance of muscle and myocardial integrity; and the suppression of inflammatory and allergic activities. Production of cortisol from the adrenal cortex is dependent upon corticotrophin (ACTH), which is secreted by the anterior pituitary. The corticotrophin releasing factor (CRF) which regulates ACTH is secreted by the hypothalamus and is responsive to cortisol levels. Physical, psychological and surgical stress and diurnal variation will affect the rate of cortisol production. Corticosteroid-binding globulin and albumin bind approximately 90% of the cortisol secreted by the adrenal cortex. Bound cortisol circulates in an available but temporarily inactive state. The physiological activity of cortisol depends upon levels of the small fraction of circulating unbound cortisol.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please properly store each component based on the instruction.
Type: Primary
Antigen: Cor
Clonality: Monoclonal
Clone: C2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Lambda
Reactivity: Multiple
Documents
Cloud-Clone Quality Guarantee
Additional Product Details
Specifications
- Size:200 µl
- Packaging:Vial
- Cross Adsorption:No
- Clonality:Monoclonal
- ImmunoPrecipitation:Yes
- Molecular Weight:362.5Da
- Purification:Protein A + Protein G affinity chromatography
- Form:Liquid
- Storage Buffer:PBS, pH 7.4, containing 0.02% NaN₃, 50% glycerol
- Isotype:IgG2b lambda
- Host:Mouse
- Concentration:1 mg/ml (for reference only)
- Cat. No.:MSPP-MA462GE21
- Reactivity:Multiple
- Shipping Temperature:Shipped on blue ice at 4 °C.
- Antibody Type:Primary
- Antigen Symbol:Cor
- Immunogen:OVA conjugated Cor
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen Synonyms:Hydrocortisone
- Antigen Name:Cortisol
- Clone:C2
- ELISA:Yes
- Storage Temperature:Store at 2...8 °C for frequent use, –20 ℃ for 24 months. Avoid repeated freeze-thaw cycles.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



