- Antibody Type:Primary
- Antigen Name:Apolipoprotein C1
- Antigen Symbol:APOC1
- Clonality:Monoclonal
- Clone:C3
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Western Blot:Yes
- Size:200 µl
- Cross Adsorption:No
- Form:Liquid
- Gene ID:11812
- Antigen Synonyms:Apo-C1|Truncated apolipoprotein C-I
- Amino Acid Number:32 - 88
- UniProtKB:P34928
- Storage Buffer:0.01 M PBS, pH 7.4, containing 0.05% Proclin-300, 50% glycerol
- Molecular Weight:37 kDa
- Storage Temperature:Store at 2...8 °C for frequent use, –20 ℃ for 24 months. Avoid repeated freeze-thaw cycles.
- Concentration:1 mg/ml (for reference only)
- Shipping Temperature:Shipped on blue ice at 4 °C.
- Immunogen:Recombinant APOC1 (Gly32~Ser88)
- Purification:Protein A + Protein G affinity chromatography
- Cat. No.:MSPP-MA252MU21
- Packaging:Vial
Monoclonal Antibody to Apolipoprotein C1 (APOC1), derived from recombinant APOC1 (Gly32~Ser88), is reactive with Mouse.
- High specificity, affinity, and purity
- Extensive validation and batch-to-batch consistency
- Diverse range of conjugated and custom options
- Comprehensive validation data and documentation
- All in stock and fast global delivery
- 100% Quality and service satisfaction guarantee
Apolipoprotein C1 is a lipoprotein with a length of 57 amino acids normally found in plasma and responsible for the activation of esterified lechitin cholesterol with an important role in the exchange of esterified cholesterol between lipoproteins and in removal of cholesterol from tissues. Its main function is inhibition of CETP, probably by altering the electric charge of HDL molecules. during fasting (like other apolipoprotein C) is found primarily within HDL, meanwhile after a meal it is found within the surface of other lipoproteins. in contrast when proteins rich in triglycerides like chylomicrons and VLDL are broken down this apoprotein is transferred again to the HDL.
Type: Primary
Antigen: APOC1
Clonality: Monoclonal
Clone: C3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Mouse