Specifications
- Antibody Type:Primary
- Antigen Name:Chemokine (C-X-C Motif) Ligand 2
- Antigen Symbol:CXCL2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western Blot:Yes
- Size:200 µl
- Cross Adsorption:No
- Form:Liquid
- Gene ID:114105
- Antigen Synonyms:GROb|GRO2|SCYB2|HSF|MIP2a|Growth Regulated Oncogene Beta|MIP2|MGSAb|Hematopoietic synergistic factor|Macrophage inflammatory protein 2-alpha|CINC2a
- Amino Acid Number:32 - 100
- UniProtKB:P30348
- Storage Buffer:0.01 M PBS, pH 7.4, containing 0.05% Proclin-300, 50% glycerol
- Molecular Weight:40.0 kDa
- Storage Temperature:Store at 2...8 °C for frequent use, –20 ℃ for 24 months. Avoid repeated freeze-thaw cycles.
- Concentration:0.5 mg/ml (for reference only)
- Shipping Temperature:Shipped on blue ice at 4 °C.
- Immunogen:Recombinant CXCL2 (Ser32~Asn100)
- Purification:Antigen-specific affinity chromatography followed by Protein A affinity chromatography
- Cat. No.:MSPP-PAB603RA1
- Packaging:Vial
Specifications
About this item
Polyclonal Antibody to Chemokine (C-X-C Motif) Ligand 2 (CXCL2), derived from recombinant CXCL2 (Ser32~Asn100), is reactive with Mouse/Rat.
- High specificity, affinity, and purity
- Extensive validation and batch-to-batch consistency
- Diverse range of conjugated and custom options
- Comprehensive validation data and documentation
- All in stock and fast global delivery
- 100% Quality and service satisfaction guarantee
Growth-regulated oncogene (GRO)beta, which belongs to the CXC chemokine family, is a potent chemoattractant for neutrophils.Chemokine (C-X-C motif) ligand 2 (CXCL2) is a small cytokine belonging to the CXC chemokine family that is also called macrophage inflammatory protein 2-alpha (MIP2-alpha), Growth-regulated protein beta (Gro-beta) and Gro oncogene-2 (Gro-2). CXCL2 is 90% identical in amino acid sequence as a related chemokine, CXCL1. This chemokine is secreted by monocytes and macrophages and is chemotactic for polymorphonuclear leukocytes and hematopoietic stem cells. The gene for CXCL2 is located on human chromosome 4 in a cluster of other CXC chemokines.CXCL2 mobilizes cells by interacting with a cell surface chemokine receptor called CXCR2.
Type: Primary
Antigen: CXCL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat