ContactUsLinkComponent
Minisart® PES Venting and Gas Filtrations
Minisart® PES Venting and Gas Filtrations
Supplier:  Sartorius
Certificates
Minisart® PES Venting and Gas Filtrations
Supplier:  Sartorius
Specifications
  • Catalog No:
  • 76474-438
  • 76474-440
  • 76474-442
  • 76474-444
  • Airflow Rate:
  • ≥1200 ml/min (0.1 bar)
  • ≥1200 ml/min (0.1 bar)
  • ≥1200 ml/min (0.1 bar)
  • ≥1200 ml/min (0.1 bar)
  • Max. Operating Pressure:
  • 4.5 bar
  • 4.5 bar
  • 4.5 bar
  • 4.5 bar
  • Max. Temperature:
  • 121 °C
  • 121 °C
  • 121 °C
  • 121 °C
Specifications