Specifications
- Antibody Type:Primary
- Antigen Name:Testis Anion Transporter 1
- Antigen Symbol:TAT1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:200 µl
- Cross Adsorption:No
- Form:Liquid
- Gene ID:116369
- Antigen Synonyms:SLC26A8|Solute Carrier Family 26 Member 8|Anion exchange transporter
- Amino Acid Number:664 - 970
- UniProtKB:Q96RN1
- Storage Buffer:0.01 M PBS, pH 7.4, containing 0.05% Proclin-300, 50% glycerol
- Molecular Weight:65 kDa
- Storage Temperature:Store at 2...8 °C for frequent use, −20 °C for 24 months. Avoid repeated freeze-thaw cycles.
- Concentration:0.5 mg/ml (for reference only)
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant TAT1(Thr664~Val970)
- Purification:Antigen-specific affinity chromatography followed by Protein A affinity chromatography
- Cat. No.:MSPP-PAE478HU1
- Packaging:Vial
Specifications
About this item
Polyclonal Antibody to Testis Anion Transporter 1 (TAT1), derived from recombinant TAT1(Thr664~Val970), is reactive with Human.
- High specificity, affinity, and purity
- Extensive validation and batch-to-batch consistency
- Diverse range of conjugated and custom options
- Comprehensive validation data and documentation
- All in stock and fast global delivery
- 100% Quality and service satisfaction guarantee
SLC26A8 is one member of a family of sulfate/anion transporter genes. Family members are well conserved in their genomic (number and size of exons) and protein (aa length among species) structures yet have markedly different tissue expression patterns. This gene is expressed primarily in spermatocytes. Two transcript variants encoding different isoforms have been found. The deduced 970-amino acid protein has a calculated molecular mass of 109 kD. SLC26A8 contains 14 putative transmembrane regions with cytoplasmic N- and C-termini. It also contains several N-glycosylation sites, 4 of which reside in the major predicted external loop. SLC26A8 shares 26% identity with pendrin (SLC26A4), and 28% identity with DRA (SLC26A3).
Type: Primary
Antigen: TAT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human