Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-VB6 Mouse Monoclonal Antibody [clone: C2]
Anti-VB6 Mouse Monoclonal Antibody [clone: C2]
Catalog # MSPP-MA916GE21
Supplier:  Cloud-Clone
Anti-VB6 Mouse Monoclonal Antibody [clone: C2]
Catalog # MSPP-MA916GE21
Supplier:  Cloud-Clone
Supplier Number:  MAA916GE21

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Vitamin B6
  • Antigen Symbol:
    VB6
  • Clonality:
    Monoclonal
  • Clone:
    C2
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Multiple
  • Size:
    200 µl
  • Cross Adsorption:
    No
  • Form:
    Liquid
  • Antigen Synonyms:
    Pyridoxine
  • Storage Buffer:
    0.01 M PBS, pH 7.4, containing 0.05% Proclin-300, 50% glycerol
  • Molecular Weight:
    169.2 Da
  • Storage Temperature:
    Store at 2...8 °C for frequent use, −20 °C for 24 months. Avoid repeated freeze-thaw cycles.
  • Concentration:
    1 mg/ml (for reference only)
  • Shipping Temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    OVA conjugated VB6
  • Purification:
    Protein A + Protein G affinity chromatography
  • Cat. No.:
    MSPP-MA916GE21
  • Packaging:
    Vial

Specifications

About this item

Monoclonal Antibody to Vitamin B6 (VB6), derived from OVA conjugated VB6, is reactive with General species.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Vitamin B6 is a water-soluble vitamin. Pyridoxine is one of the compounds that can be called vitamin B6, along with pyridoxal and pyridoxamine. Its hydrochloride salt pyridoxine hydrochloride is often used.  Vitamin B6 assists in the balancing of sodium and potassium as well as promoting red blood cell production. It is linked to cardiovascular health by decreasing the formation of homocysteine. Pyridoxine may help balance hormonal changes in women and aid the immune system. It is required for the production of the monoamine neurotransmitters serotonin, dopamine, norepinephrine and epinephrine, as it is the precursor to pyridoxal phosphate: cofactor for the enzyme aromatic amino acid decarboxylase. This enzyme is responsible for converting the precursors 5-hydroxytryptophan (5-HTP) into serotonin and melatonin, and levodopa (L-DOPA) into dopamine, noradrenaline and adrenaline.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: VB6
Clonality: Monoclonal
Clone: C2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: General species