Specifications
- Antibody Type:Primary
- Antigen Name:Ribonuclease A4
- Antigen Symbol:RNASE4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Western Blot:Yes
- Size:200 µl
- Cross Adsorption:No
- Form:Liquid
- Gene ID:6038
- Antigen Synonyms:Ribonuclease,RNase A Family 4|Rnase-A4|RNS4
- Amino Acid Number:30 - 147
- UniProtKB:P34096
- Storage Buffer:0.01 M PBS, pH 7.4, containing 0.05% Proclin-300, 50% glycerol
- Molecular Weight:18 kDa
- Storage Temperature:Store at 2...8 °C for frequent use, −20 °C for 24 months. Avoid repeated freeze-thaw cycles.
- Concentration:0.5 mg/ml (for reference only)
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant RNASE4(Asp30~Gly147)
- Purification:Antigen-specific affinity chromatography followed by Protein A affinity chromatography
- Cat. No.:MSPP-PAD190HU1
- Packaging:Vial
Specifications
About this item
Polyclonal Antibody to Ribonuclease A4 (RNASE4), derived from recombinant RNASE4(Asp30~Gly147), is reactive with Human/Rat.
- High specificity, affinity, and purity
- Extensive validation and batch-to-batch consistency
- Diverse range of conjugated and custom options
- Comprehensive validation data and documentation
- All in stock and fast global delivery
- 100% Quality and service satisfaction guarantee
Rosenberg and Dyer (1995) referred to the protein as ribonuclease 4 (RNase4). By PCR analysis of human/hamster somatic cell hybrids, they determined that the gene encoding the protein resides in a single exon found on human chromosome 14.By Northern analysis, mRNA for the gene was detected in human pancreas, lung, skeletal muscle, heart, kidney, and placenta, but not brain; liver represented the most abundant source. The authors noted that the mRNA encoding this ribonuclease is approximately 2 kb in length, or approximately twice as long as the mRNAs encoding other members of this gene family. They also detected a larger (approximately 2.4 kb), second transcript in hepatic, pancreatic and renal tissues. At the time of creation of this entry, 1 other protein had the name ribonuclease 4: RNS4, which is located on chromosome 1.
Type: Primary
Antigen: RNASE4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Rat