Specifications
- Antibody Type:Primary
- Antigen Name:Epithelial Cell Adhesion Molecule
- Antigen Symbol:EPCAM
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Chicken
- Western Blot:Yes
- Size:200 µl
- Cross Adsorption:No
- Form:Liquid
- Gene ID:421292
- Antigen Synonyms:CD326|Epithelial glycoprotein|TACSTD1|MIC18|M4S1|KSA|Adenocarcinoma-associated antigen|TROP1|ESA|Tumor Associated Calcium Signal Transducer 1|HEGP2|EGP
- Amino Acid Number:21 - 260
- UniProtKB:Q5F381
- Storage Buffer:PBS, pH 7.4, containing 0.02% NaN₃, 50% glycerol
- Molecular Weight:36 kDa
- Storage Temperature:Store at 2...8 °C for frequent use, −20 °C for 24 months. Avoid repeated freeze-thaw cycles.
- Concentration:0.79 mg/ml (for reference only)
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant EPCAM(Asp21~Gly260)
- Purification:Antigen-specific affinity chromatography followed by Protein A affinity chromatography
- Cat. No.:MSPP-PAB283GA1
- Packaging:Vial
Specifications
About this item
Polyclonal Antibody to Epithelial Cell Adhesion Molecule (EPCAM), derived from recombinant EPCAM(Asp21~Gly260), is reactive with Chicken.
- High specificity, affinity, and purity
- Extensive validation and batch-to-batch consistency
- Diverse range of conjugated and custom options
- Comprehensive validation data and documentation
- All in stock and fast global delivery
- 100% Quality and service satisfaction guarantee
The epithelial cell adhesion molecule (EpCAM, CD326) is a glycoprotein of ~40 kd that was originally identified as a marker for carcinoma, attributable to its high expression on rapidly proliferating tumors of epithelial origin. Normal epithelia express EpCAM at a variable but generally lower level than carcinoma cells. In early studies, EpCAM was proposed to be a cell-cell adhesion molecule.,However, recent insights revealed a more versatile role for EpCAM that is not limited only to cell adhesion but includes diverse processes such as signaling, cell migration, proliferation, and differentiation. Cell surface expression of EpCAM may actually prevent cell-cell adhesion.
Type: Primary
Antigen: EPCAM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Chicken