Specifications
- Antibody Type:Primary
- Antigen Name:Adenylate Cyclase 7
- Antigen Symbol:ADCY7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:200 µl
- Cross Adsorption:No
- Form:Liquid
- Gene ID:11513
- Antigen Synonyms:ATP pyrophosphate-lyase 7|AC7|Adenylate cyclase type VII
- Amino Acid Number:309 - 491
- UniProtKB:P51829
- Storage Buffer:0.01 M PBS, pH 7.4, containing 0.05% Proclin-300, 50% glycerol
- Molecular Weight:27 kDa
- Storage Temperature:Store at 2...8 °C for frequent use, −20 °C for 24 months. Avoid repeated freeze-thaw cycles.
- Concentration:0.5 mg/ml (for reference only)
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant ADCY7(Leu309~Glu491)
- Purification:Antigen-specific affinity chromatography followed by Protein A affinity chromatography
- Cat. No.:MSPP-PAB419MU1
- Packaging:Vial
Specifications
About this item
Polyclonal Antibody to Adenylate Cyclase 7 (ADCY7), derived from recombinant ADCY7(Leu309~Glu491), is reactive with Human/Mouse.
- High specificity, affinity, and purity
- Extensive validation and batch-to-batch consistency
- Diverse range of conjugated and custom options
- Comprehensive validation data and documentation
- All in stock and fast global delivery
- 100% Quality and service satisfaction guarantee
Adenylate cyclase type 7 is a membrane-bound adenylate cyclase that catalyses the formation of cyclic AMP from ATP and is inhibitable by calcium. The product of this gene is a member of the adenylyl cyclase class-4/guanylyl cyclase enzyme family that is characterized by the presence of twelve membrane-spanning domains in its sequences.The deduced protein contains 12 membrane-spanning domains, a characteristic of the adenylyl cyclase enzyme family. RNase protection assays indicated that ADCY7 is the major form of adenylyl cyclase in human platelets. Northern blot analysis detected a 6.7-kb transcript that was abundant in liver. Of brain areas studied, the transcript was most abundant in the caudate and cerebellum and present at a slightly lower level in hippocampus.
Type: Primary
Antigen: ADCY7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse