- Antibody Type:Primary
- Antigen Name:Chondroitin Sulfate
- Antigen Symbol:CS
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Multiple
- Size:200 µl
- Cross Adsorption:No
- Form:Liquid
- Storage Buffer:0.01 M PBS, pH 7.4, containing 0.05% Proclin-300, 50% glycerol
- Storage Temperature:Store at 2...8 °C for frequent use, −20 °C for 24 months. Avoid repeated freeze-thaw cycles.
- Concentration:0.5 mg/ml (for reference only)
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:BSA conjugated CS
- Purification:Antigen-specific affinity chromatography followed by Protein A affinity chromatography
- Cat. No.:MSPP-PAA723GE1
- Packaging:Vial
Polyclonal Antibody to Chondroitin Sulfate (CS), derived from BSA conjugated CS, is reactive with General species.
- High specificity, affinity, and purity
- Extensive validation and batch-to-batch consistency
- Diverse range of conjugated and custom options
- Comprehensive validation data and documentation
- All in stock and fast global delivery
- 100% Quality and service satisfaction guarantee
Chondroitin sulfate (CS) is a major microenvironmental molecule in the CNS, and there have been few reports about its neuroprotective activity. One of several classes of sulfated glycosaminoglycans that is a major constituent in various connective tissues, especially in the ground substance of blood vessels, bone, and cartilage.,Chondroitin sulfate is a sulfated glycosaminoglycan (GAG) composed of a chain of alternating sugars (N-acetylgalactosamine and glucuronic acid). It is usually found attached to proteins as part of a proteoglycan. A chondroitin chain can have over 100 individual sugars, each of which can be sulfated in variable positions and quantities. Understanding the functions of such diversity in chondroitin sulfate and related glycosaminoglycans is a major goal of glycobiology. Chondroitin sulfate is an important structural component of cartilage and provides much of its resistance to compression .
Type: Primary
Antigen: CS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: General species