Specifications
- Antibody Type:Primary
- Antigen Name:Calmodulin Like Protein 3
- Antigen Symbol:CALML3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Western Blot:Yes
- Size:200 µl
- Cross Adsorption:No
- Form:Liquid
- Gene ID:810
- Antigen Synonyms:Calmodulin-related protein NB-1|CLP|CaM-like protein
- Amino Acid Number:1 - 149
- UniProtKB:P27482
- Storage Buffer:0.01 M PBS, pH 7.4, containing 0.05% Proclin-300, 50% glycerol
- Molecular Weight:21 kDa
- Storage Temperature:Store at 2...8 °C for frequent use, −20 °C for 24 months. Avoid repeated freeze-thaw cycles.
- Concentration:0.5 mg/ml (for reference only)
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant CALML3(Met1~Lys149)
- Purification:Antigen-specific affinity chromatography followed by Protein A affinity chromatography
- Cat. No.:MSPP-PAF898HU1
- Packaging:Vial
Specifications
About this item
Polyclonal Antibody to Calmodulin Like Protein 3 (CALML3), derived from recombinant CALML3(Met1~Lys149), is reactive with Human/Rat.
- High specificity, affinity, and purity
- Extensive validation and batch-to-batch consistency
- Diverse range of conjugated and custom options
- Comprehensive validation data and documentation
- All in stock and fast global delivery
- 100% Quality and service satisfaction guarantee
CALML3, the deduced 148-amino acid protein contains 4 helix-loop-helix Ca(2+)-binding motifs and shares 85% identity with vertebrate calmodulin. Northern blot analysis detected no expression in fibroblast, teratoma, or erythroleukemia cell lines. CLP was expressed exclusively in the epithelium of the tissues surveyed and was most abundant in thyroid, breast, prostate, kidney, and skin. CLP expression appeared to increase in stratified epithelium during differentiation, as illustrated in skin, where CLP staining intensified from the basal through the spinous to the granular layers.Significant differences between the electric surface potentials at the target protein-binding regions of CLP and calmodulin suggested that the ranges of CLP and calmodulin target proteins do not fully overlap.
Type: Primary
Antigen: CALML3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Rat