- Antibody Type:Primary
- Antigen Name:Urocortin
- Antigen Symbol:UCN
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Mouse
- Western Blot:Yes
- Size:200 µl
- Cross Adsorption:No
- Form:Liquid
- Gene ID:22226
- Antigen Synonyms:UI|UCN1|Urocortin 1|UROC
- Amino Acid Number:34 - 120
- UniProtKB:P81615
- Storage Buffer:0.01 M PBS, pH 7.4, containing 0.05% Proclin-300, 50% glycerol
- Molecular Weight:40 kDa
- Storage Temperature:Store at 2...8 °C for frequent use, −20 °C for 24 months. Avoid repeated freeze-thaw cycles.
- Concentration:0.5 mg/ml (for reference only)
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant UCN(Thr34~Val120)
- Purification:Antigen-specific affinity chromatography followed by Protein A affinity chromatography
- Cat. No.:MSPP-PAA231MU1
- Packaging:Vial
Polyclonal Antibody to Urocortin (UCN), derived from recombinant UCN(Thr34~Val120), is reactive with Mouse.
- High specificity, affinity, and purity
- Extensive validation and batch-to-batch consistency
- Diverse range of conjugated and custom options
- Comprehensive validation data and documentation
- All in stock and fast global delivery
- 100% Quality and service satisfaction guarantee
Urocortin is a member of the sauvagine/corticotropin-releasing factor/urotensin I family. It is structurally related to the corticotropin-releasing factor (CRF) gene and the encoded product is an endogenous ligand for CRF type 2 receptors. In the brain it may be responsible for the effects of stress on appetite. In spite of the gene family name similarity, the product of this gene has no sequence similarity to urotensin II. Urocortin is a potent anorexigenic peptide of 40 amino acids that induces fed-like motor activity when administered centrally or peripherally in fasted animals. Urocortin belongs to the corticotropin-releasing factor (CRF) family that includes CRF, urotensin I, sauvagine, urocortin II and urocortin III. Urocortin is also a potent and long-lasting hypotensive agent and increases coronary blood flow.
Type: Primary
Antigen: UCN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse