To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Sodium/potassium Transporting ATPase Subunit Beta-3
- Antigen Symbol:ATP1b3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Mouse
- Western Blot:Yes
- Size:200 µl
- Cross Adsorption:No
- Form:Liquid
- Gene ID:11933
- Antigen Synonyms:Na+/K+ Transporting Beta 3 Polypeptide|ATPB-3|ATP1-B3|CD298|ATPase
- Amino Acid Number:61 - 277
- UniProtKB:P97370
- Storage Buffer:PBS, pH 7.4, containing 0.02% NaN₃, 50% glycerol
- Molecular Weight:31 kDa
- Storage Temperature:Store at 2...8 °C for frequent use, −20 °C for 24 months. Avoid repeated freeze-thaw cycles.
- Concentration:1 mg/ml (for reference only)
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant ATP1b3(Met61~Arg277)
- Purification:Antigen-specific affinity chromatography followed by Protein A affinity chromatography
- Cat. No.:MSPP-PAA712MU1
- Packaging:Vial
Specifications
About this item
Polyclonal Antibody to Sodium/potassium Transporting ATPase Subunit Beta-3 (ATP1b3), derived from recombinant ATP1b3(Met61~Arg277), is reactive with Mouse.
- High specificity, affinity, and purity
- Extensive validation and batch-to-batch consistency
- Diverse range of conjugated and custom options
- Comprehensive validation data and documentation
- All in stock and fast global delivery
- 100% Quality and service satisfaction guarantee
Sodium/potassium-transporting ATPase subunit beta-3 belongs to the family of Na /K and H /K ATPases beta chain proteins, and to the subfamily of Na+/K+-ATPases. Na+/K+-ATPase is an integral membrane protein responsible for establishing and maintaining the electrochemical gradients of Na and K ions across the plasma membrane. These gradients are essential for osmoregulation, for sodium-coupled transport of a variety of organic and inorganic molecules, and for electrical excitability of nerve and muscle. This enzyme is composed of two subunits, a large catalytic subunit (alpha) and a smaller glycoprotein subunit (beta). The beta subunit regulates, through assembly of alpha/beta heterodimers, the number of sodium pumps transported to the plasma membrane.
Type: Primary
Antigen: ATP1b3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse