Order Entry
Puerto Rico
ContactUsLinkComponent
Sodium Citrate Buffer, 0.5 M, pH 4.5, 250 ml, RNase/DNase Tested, Sterile
Sodium Citrate Buffer, 0.5 M, pH 4.5, 250 ml, RNase/DNase Tested, Sterile
Catalog # 77510-020
Supplier:  Teknova
CAS Number:  
Sodium Citrate Buffer, 0.5 M, pH 4.5, 250 ml, RNase/DNase Tested, Sterile
Catalog # 77510-020
Supplier:  Teknova
Supplier Number:  S8511
CAS Number:  

Specifications

  • Description:
    0.5 M sodium citrate
  • Size:
    250 ml
  • Sterility:
    Sterile
  • Storage Temperature:
    RT
  • pH buffer type:
    Citrate
  • Cat. No.:
    77510-020
  • pH:
    4.50 ±0.04

Specifications

About this item