SD-169 ≥97% (by HPLC), Calbiochem®, Sigma-Aldrich®
82603-188
:
- Pack type:Plastic ampoule
- Size:10 mg
- Cat. No.:82603-188
- Supplier no.:506158-10MG
:
:160.17 g/mol
:M-Clarity™ quality level = MQ100
:Refrigerator
:1670-87-7
:C₉H₈N₂O
:MFCD07779486