IQ-1 ≥95% (by HPLC), Calbiochem®, Sigma-Aldrich®
Supplier: MilliporeSigma
Certificates
About this item
CAS Number:331001-62-8
:
Formula:C₂₁H₂₂N₄O₂
MDL Number:MFCD00354369
:362.43 g/mol
:M-Clarity™ quality level = MQ100
Storage Temperature:Refrigerator