DL-Sulforaphane ≥98% (by UPLC), Calbiochem®, Sigma-Aldrich®
Catalog # 80512-426
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Plastic ampule
- Size:25 mg
- Cat. No.:80512-426
- Supplier no.:574215-25MG
Specifications
About this item
:
:177.3 g/mol
Storage Temperature:Freezer
CAS Number:4478-93-7
Formula:C₆H₁₁NOS₂
MDL Number:MFCD00198068