Order Entry
Puerto Rico
ContactUsLinkComponent
 
Health
Simvastatin ≥98% (by HPLC), Calbiochem®, Sigma-Aldrich®
Catalog # 80503-392
Supplier:  MilliporeSigma
CAS Number:  
Health
Simvastatin ≥98% (by HPLC), Calbiochem®, Sigma-Aldrich®
Catalog # 80503-392
Supplier:  MilliporeSigma
Supplier Number:  567020-50MG
CAS Number:  

Specifications

  • Pack type:
    Plastic ampule
  • Size:
    50 mg
  • Cat. No.:
    80503-392
  • Supplier no.:
    567020-50MG

Specifications

About this item

 :
 :418.57 g/mol
 :GHS08
Storage Temperature:Refrigerator
CAS Number:79902-63-9
Formula:C25H38O5
MDL Number:MFCD00072007