Order Entry
Puerto Rico
ContactUsLinkComponent
L-Phenylalanine ≥98.5% (by titrimetric analysis on dry substance), Calbiochem®, Millipore®
L-Phenylalanine ≥98.5% (by titrimetric analysis on dry substance), Calbiochem®, Millipore®
Catalog # 80058-546
Supplier:  MilliporeSigma
CAS Number:  
L-Phenylalanine ≥98.5% (by titrimetric analysis on dry substance), Calbiochem®, Millipore®
Catalog # 80058-546
Supplier:  MilliporeSigma
Supplier Number:  5202-100GM
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Size:
    100 g
  • Cat. No.:
    80058-546
  • Supplier no.:
    5202-100GM

Specifications

About this item

 :
 :165.19 g/mol
Melting Pt:275-283 °C
Density:1.34 g/cm3 (20 °C)
Storage Temperature:Ambient
CAS Number:63-91-2
Formula:C₉H₁₁NO₂
MDL Number:MFCD00064227
Melting Pt:275…283 °C