L-Methionine ≥98.5% (by assay), Calbiochem®, Millipore®
Catalog # 80058-348
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Glass bottle
- Size:100 g
- Cat. No.:80058-348
- Supplier no.:4500-100GM
Specifications
About this item
:
:149.21 g/mol
Storage Temperature:Ambient
CAS Number:63-68-3
Formula:C₅H₁₁NO₂S
MDL Number:MFCD00063097