L(+)-Lysine monohydrochloride ≥98% (by titrimetric analysis on dry substance), Calbiochem®, Millipore®
Supplier: MilliporeSigma
Certificates
About this item
CAS Number:657-27-2
:
Formula:H₂N(CH₂)₄CH(NH₂)CO₂H·HCl
MDL Number:MFCD00064564
:182.65 g/mol
Storage Temperature:Ambient