Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
L(+)-Isoleucine
L(+)-Isoleucine ≥99% (by TLC), Calbiochem®, Millipore®
L(+)-Isoleucine ≥99% (by TLC), Calbiochem®, Millipore®
80058-284
:
MilliporeSigma
:
L(+)-Isoleucine ≥99% (by TLC), Calbiochem®, Millipore®
80058-284
:
MilliporeSigma
:
4160-100GM
:
Pack type:
Glass bottle
Size:
100 g
Cat. No.:
80058-284
Supplier no.:
4160-100GM
:
:
131.17 g/mol
:
Ambient
:
73-32-5
:
H₃CCH₂CH(CH₃)CH(NH₂)COOH
:
MFCD00064222