Order Entry
Puerto Rico
ContactUsLinkComponent
5-Hydroxy-L-tryptophan ≥98% (by HPLC), Calbiochem®, Millipore®
5-Hydroxy-L-tryptophan ≥98% (by HPLC), Calbiochem®, Millipore®
Catalog # 80056-726
Supplier:  MilliporeSigma
CAS Number:  
5-Hydroxy-L-tryptophan ≥98% (by HPLC), Calbiochem®, Millipore®
Catalog # 80056-726
Supplier:  MilliporeSigma
Supplier Number:  399698-25GM
CAS Number:  

Specifications

  • Pack type:
    Plastic ampule
  • Size:
    25 g
  • Cat. No.:
    80056-726
  • Supplier no.:
    399698-25GM

Specifications

About this item

 :
 :220.23 g/mol
Storage Temperature:Refrigerator
UN:2811
ADR:6.1,III
 :GHS06
CAS Number:4350-09-8
Formula:C₁₁H₁₂N₂O₃
MDL Number:MFCD00064341