Order Entry
Puerto Rico
ContactUsLinkComponent
Fura 2-AM in solution ≥95% (by HPLC), Calbiochem®, Sigma-Aldrich®
Fura 2-AM in solution ≥95% (by HPLC), Calbiochem®, Sigma-Aldrich®
Catalog # 80051-792
Supplier:  MilliporeSigma
CAS Number:  
Fura 2-AM in solution ≥95% (by HPLC), Calbiochem®, Sigma-Aldrich®
Catalog # 80051-792
Supplier:  MilliporeSigma
Supplier Number:  344906-1ML
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Plastic ampule
  • Size:
    1 ml
  • Cat. No.:
    80051-792
  • Supplier no.:
    344906-1ML

Specifications

About this item

 :
 :1001.857 g/mol
Storage Temperature:Freezer
UN:1993
CAS Number:108964-32-5
Formula:C₄₄H₄₇N₃O₂₄
MDL Number:MFCD00036976