Order Entry
Puerto Rico
ContactUsLinkComponent
 
5-Bromo-4-chloro-3-indolyl phosphate disodium salt (BCIP-Na) ≥99% (by HPLC), Calbiochem®, Sigma-Aldrich®
Catalog # EM203788-1GM
Supplier:  MilliporeSigma
CAS Number:  
5-Bromo-4-chloro-3-indolyl phosphate disodium salt (BCIP-Na) ≥99% (by HPLC), Calbiochem®, Sigma-Aldrich®
Catalog # EM203788-1GM
Supplier:  MilliporeSigma
Supplier Number:  203788-1GM
CAS Number:  

Specifications

  • Pack type:
    Plastic ampule
  • Size:
    1 g
  • Cat. No.:
    EM203788-1GM
  • Supplier no.:
    203788-1GM

Specifications

About this item

 :
 :370.43 g/mol
Storage Temperature:Freezer
CAS Number:102185-33-1
Formula:C₈H₄BrClNNa₂O₄P
MDL Number:MFCD00036757