Order Entry
Puerto Rico
ContactUsLinkComponent
 
L-Alanine ≥98% (by titrimetric analysis on dry substance), Calbiochem®, Millipore®
Catalog # 80057-774
Supplier:  MilliporeSigma
CAS Number:  
L-Alanine ≥98% (by titrimetric analysis on dry substance), Calbiochem®, Millipore®
Catalog # 80057-774
Supplier:  MilliporeSigma
Supplier Number:  1250-100GM
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Size:
    100 g
  • Cat. No.:
    80057-774
  • Supplier no.:
    1250-100GM

Specifications

About this item

 :
 :89.09 g/mol
Storage Temperature:Ambient
CAS Number:56-41-7
Formula:C₃H₇NO₂
MDL Number:MFCD00064410