Order Entry
Puerto Rico
ContactUsLinkComponent
8-(4-Chlorophenylthio)adenosine-3',5'-cyclic monophosphate sodium salt ≥98% (by HPLC), Calbiochem®, Sigma-Aldrich®
8-(4-Chlorophenylthio)adenosine-3',5'-cyclic monophosphate sodium salt ≥98% (by HPLC), Calbiochem®, Sigma-Aldrich®
Catalog # 80050-092
Supplier:  MilliporeSigma
CAS Number:  
8-(4-Chlorophenylthio)adenosine-3',5'-cyclic monophosphate sodium salt ≥98% (by HPLC), Calbiochem®, Sigma-Aldrich®
Catalog # 80050-092
Supplier:  MilliporeSigma
Supplier Number:  116812-10MG
CAS Number:  

Specifications

  • Pack type:
    Plastic ampule
  • Size:
    10 mg
  • Cat. No.:
    80050-092
  • Supplier no.:
    116812-10MG

Specifications

About this item

 :
 :493.791 g/mol
Storage Temperature:Freezer
CAS Number:93882-12-3
Formula:C₁₆H₁₄ClN₅NaO₆PS
MDL Number:MFCD00057683