Order Entry
Puerto Rico
ContactUsLinkComponent
D-(-)-Mannitol for microbiology, Millipore®
D-(-)-Mannitol for microbiology, Millipore®
Catalog # EM1.05982.0500
Supplier:  MilliporeSigma
CAS Number:  
D-(-)-Mannitol for microbiology, Millipore®
Catalog # EM1.05982.0500
Supplier:  MilliporeSigma
Supplier Number:  1.05982.0500
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    500 g
  • Cat. No.:
    EM1.05982.0500
  • Supplier no.:
    1.05982.0500

Specifications

About this item

 :
 :182.17 g/mol
Boiling Pt:290-295 °C/4 hPa
Boiling Pt:290…295 °C/4 hPa
Melting Pt:167-170 °C (lit.)
Melting Pt:167…170 °C (lit.)
Storage Temperature:Ambient
CAS Number:69-65-8
Formula:HOH₂C(CH(OH))₄CH₂OH
MDL Number:MFCD00064287