Orders LinkContactUsLinkComponent
Cold Transport Containers with Packaging
Cold Transport Containers with Packaging
Supplier:   SARSTEDT INC
Cold Transport Containers with Packaging
Supplier:  SARSTEDT INC
Certificates
Cold Transport Containers with Packaging
Cold Transport Containers with Packaging
2 Options Available Below
Product Details & Documents
SARSTEDT offers sample transport systems and secondary containers for a wide variety of transport requirements.
  • Mailing packaging for the secure cold transport of medical and biological examination material
  • No absorbent liner
  • Suitable for 2×10 ml sample tubes, e.g. 9 ml S-Monovette® or one tube with max. 16.5 mm diameter and a max. total length of 107 mm
  • Mailing container is filled with coolant and is frozen lying down to at least –12 °C before transport
  • Customized information can be printed on request
  • Not suitable for samples that must not be frozen as an unfrozen sample would freeze in the deep-freeze container
SARSTEDT offers standard and specialty transport systems for specimens.
Product Family Options
Product Information
  • Dimensions
    Availability
    Price
  • 77589-938
    Cold transport container with packaging   
    Cold transport container with packaging   
    140×71×168 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 1
    Pack of 1 - $11.28
    $11.28
    Log in for availability
    Case of 36
    Case of 36 - $406.08
    $406.08
    Pack of 1
       $11.28
    Case of 36
       $406.08
    Supplier Number: 95.1011
  • 77589-942
    Cold transport container with packaging, labelled in English, P 650, ADR base shape
    Cold transport container with packaging, labelled in English, P 650, ADR base shape
    140×71×170 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 1
    Pack of 1 - $96.95
    $96.95
    Log in for availability
    Case of 6
    Case of 6 - $581.70
    $581.70
    Pack of 1
       $96.95
    Case of 6
       $581.70
    Supplier Number: 95.1123