Mailing Containers, Round
New Product
Sarstedt offers sample transport systems and secondary containers for a wide variety of transport requirements.
- Corresponds to the requirement: P 650, ADR
- Closure: Without
- Made of PP
- The tight seal with a pressure difference of 95 kPa makes them ideal as secondary packaging as per packing instruction
- Autoclavable at 121 °C
- Available with and without an absorbent liner
- Available with customized printed text on request
Mailing containers with screw cap and it is a secure container for blood, urine and stool sample tubes.