Order Entry
Puerto Rico
ContactUsLinkComponent
Treston Cable Tidy
Treston Cable Tidy
  77575-670
 :  Treston
New Product
undefined
Treston Cable Tidy
  77575-670
 :  Treston
 :  TCT-49
New Product

 

  • Description:
    Treston cable tidy
  • Cat. No.:
    77575-670
  • W×D×H:
    800×62×100 mm
  • Weight:
    1.4 kg

 

 

  • Color: Light gray
  • RAL code: RAL7035