RecombiMAb Anti-PD-1 Mouse Recombinant Antibody [clone: RMP1-14-CP151]
Supplier: Bio X Cell
New Product
Certificates
About this item
Type: Primary
Antigen: PD-1
Clonality: Recombinant
Clone: RMP1-14-CP151
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Mouse
Specifications
- Catalog No:
- 77580-268
- 77580-266
- 77580-264
- 77580-262
- 77580-260
- Antigen Symbol:
- PD-1
- PD-1
- PD-1
- PD-1
- PD-1
- Antibody Type:
- Primary
- Primary
- Primary
- Primary
- Primary
- Clonality:
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Clone:
- RMP1-14-CP151
- RMP1-14-CP151
- RMP1-14-CP151
- RMP1-14-CP151
- RMP1-14-CP151
- Host:
- Mouse
- Mouse
- Mouse
- Mouse
- Mouse
- Tested Applications:
- in vivo blocking of PD-1/PD-L signaling*
*Reported for the original rat IgG2a RMP1-14 antibody - in vivo blocking of PD-1/PD-L signaling*
*Reported for the original rat IgG2a RMP1-14 antibody - in vivo blocking of PD-1/PD-L signaling*
*Reported for the original rat IgG2a RMP1-14 antibody - in vivo blocking of PD-1/PD-L signaling*
*Reported for the original rat IgG2a RMP1-14 antibody - in vivo blocking of PD-1/PD-L signaling*
*Reported for the original rat IgG2a RMP1-14 antibody